Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331926.1 | NP_001318975.1 | 29 - 30 | 3975 | 90 | |
NM_001331927.1 | NP_001320611.1 | 28 - 29 | 3824 | 90 | ||
NM_100952.4 | NP_563877.1 | 30 - 31 | 3809 | 90 | ||
GenBank mRNA | AF312027.1 | - | 27 - 28 | 3515 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |