Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331945.1 | NP_001318977.1 | 1 - 2 | 676 | 67 | |
NM_001331946.1 | NP_001320672.1 | 2 - 3 | 1084 | 67 | ||
GenBank mRNA | AK221596.1 | - | 3 - 4 | 972 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |