Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331947.1 | NP_001320915.1 | 2 - 3 | 328 | 126 | |
NM_001331948.1 | NP_001320916.1 | 3 - 4 | 320 | 126 | ||
NM_100975.3 | NP_563882.1 | 2 - 3 | 336 | 126 | ||
GenBank mRNA | AF369565.1 | - | 2 - 3 | 243 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |