Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331960.1 | NP_001321706.1 | 10 - 11 | 1508 | 61 | |
NM_100986.4 | NP_172580.2 | 11 - 12 | 1992 | 61 | ||
GenBank mRNA | AF399923.1 | - | 11 - 12 | 1781 | 61 | |
AK227100.1 | - | 11 - 12 | 1793 | 61 | ||
AY091019.1 | - | 11 - 12 | 1843 | 61 | ||
AY117318.1 | - | 11 - 12 | 1565 | 61 | ||
FJ708632.1 | - | 11 - 12 | 1565 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |