Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345226.1 | NP_001331716.1 | 1 - 2 | 645 | 89 | |
NM_125079.3 | NP_200507.1 | 1 - 2 | 653 | 89 | ||
GenBank mRNA | AF303979.1 | - | 1 - 2 | 606 | 89 | |
BT033021.1 | - | 1 - 2 | 606 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Arabidopsis thaliana Probe spans exons |