Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345226.1 | NP_001331716.1 | 2 - 3 | 777 | 140 | |
NM_125079.3 | NP_200507.1 | 2 - 3 | 785 | 140 | ||
GenBank mRNA | AF303979.1 | - | 2 - 3 | 738 | 140 | |
BT033021.1 | - | 2 - 3 | 738 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |