Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT5G57300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085291.3 | NP_001078760.1 | 7 - 8 | 824 | 81 | |
NM_001203629.1 | NP_001190558.1 | 7 - 8 | 817 | 81 | ||
NM_001345255.1 | NP_001332423.1 | 6 - 7 | 696 | 81 | ||
NM_125112.5 | NP_200540.1 | 7 - 8 | 774 | 81 | ||
GenBank mRNA | AY050939.1 | - | 7 - 8 | 773 | 81 | |
AY091173.1 | - | 6 - 7 | 647 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |