Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RR18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125193.4 | NP_200616.4 | 5 - 6 | 1856 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |