Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ftsh9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125277.4 | NP_568892.1 | 1 - 2 | 1082 | 61 | |
GenBank mRNA | AY059856.1 | - | 1 - 2 | 889 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |