Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345330.1 | NP_001318835.1 | 3 - 4 | 1471 | 115 | |
NM_001345331.1 | NP_001332082.1 | 4 - 5 | 916 | 115 | ||
NM_001345332.1 | NP_001332083.1 | 3 - 4 | 868 | 115 | ||
NM_125284.4 | NP_200702.2 | 2 - 3 | 946 | 115 | ||
GenBank mRNA | BT015092.1 | - | 2 - 3 | 617 | 115 | |
BT015906.1 | - | 2 - 3 | 544 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |