Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORP3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125333.5 | NP_200750.1 | 7 - 8 | 960 | 72 | |
GenBank mRNA | AY054473.1 | - | 7 - 8 | 913 | 72 | |
BT000115.1 | - | 7 - 8 | 796 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |