Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345367.1 | NP_001329954.1 | 6 - 7 | 1150 | 68 | |
NM_001345368.1 | NP_001329955.1 | 5 - 6 | 1621 | 68 | ||
NM_125363.3 | NP_568912.2 | 5 - 6 | 1650 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |