Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345367.1 | NP_001329954.1 | 11 - 12 | 1879 | 95 | |
NM_001345368.1 | NP_001329955.1 | 10 - 11 | 2350 | 95 | ||
NM_125363.3 | NP_568912.2 | 10 - 11 | 2379 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |