Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125462.4 | NP_200877.1 | 2 - 3 | 1154 | 89 | |
GenBank mRNA | AF188994.1 | - | 2 - 3 | 905 | 89 | |
AF233592.1 | - | 2 - 3 | 588 | 89 | ||
AK229564.1 | - | 2 - 3 | 1112 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |