Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345435.1 | NP_001330939.1 | 4 - 5 | 1064 | 63 | |
NM_125485.4 | NP_568930.1 | 4 - 5 | 1048 | 63 | ||
GenBank mRNA | AY094402.1 | - | 4 - 5 | 749 | 63 | |
BT001119.1 | - | 4 - 5 | 642 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |