Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125531.4 | NP_200946.1 | 1 - 2 | 711 | 113 | |
GenBank mRNA | AB041530.1 | - | 1 - 2 | 144 | 113 | |
AF272039.1 | - | 1 - 2 | 531 | 113 | ||
AY094393.1 | - | 1 - 2 | 586 | 113 | ||
BT005816.1 | - | 1 - 2 | 134 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |