Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037043.1 | NP_001032120.1 | 1 - 2 | 316 | 76 | |
NM_125551.4 | NP_200966.2 | 1 - 2 | 366 | 76 | ||
GenBank mRNA | AY099694.1 | - | 1 - 2 | 314 | 76 | |
AY128873.1 | - | 1 - 2 | 257 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |