Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALATS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198257.1 | NP_001185186.2 | 14 - 15 | 2061 | 82 | |
NM_103905.4 | NP_175439.2 | 14 - 15 | 2151 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |