Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALATS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198257.1 | NP_001185186.2 | 7 - 8 | 1371 | 81 | |
NM_103905.4 | NP_175439.2 | 7 - 8 | 1461 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |