Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333427.1 | NP_001319189.1 | 18 - 19 | 3402 | 127 | |
NM_001333428.1 | NP_001321239.1 | 18 - 19 | 3335 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |