Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULTR2;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121056.3 | NP_196580.1 | 4 - 5 | 1332 | 96 | |
GenBank mRNA | AB003591.1 | - | 4 - 5 | 1191 | 96 | |
AY062546.1 | - | 4 - 5 | 1205 | 96 | ||
AY093335.1 | - | 4 - 5 | 1124 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |