Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANNAT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121061.3 | NP_196585.1 | 1 - 2 | 156 | 88 | |
GenBank mRNA | BT029226.1 | - | 1 - 2 | 69 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |