Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMB3010 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203345.1 | NP_001190274.1 | 2 - 3 | 169 | 91 | |
NM_121074.5 | NP_196598.1 | 2 - 3 | 189 | 91 | ||
GenBank mRNA | AY045594.1 | - | 2 - 3 | 162 | 91 | |
AY057634.1 | - | 2 - 3 | 162 | 91 | ||
AY093783.1 | - | 1 - 2 | 77 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |