Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121119.3 | NP_568240.1 | 3 - 4 | 358 | 118 | |
GenBank mRNA | AK117573.1 | - | 3 - 4 | 301 | 118 | |
BT005033.1 | - | 2 - 3 | 222 | 118 | ||
U67398.1 | - | 3 - 4 | 260 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |