Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343159.1 | NP_001329978.1 | 3 - 4 | 770 | 79 | |
NM_121154.4 | NP_196677.1 | 3 - 4 | 770 | 79 | ||
GenBank mRNA | BT004028.1 | - | 3 - 4 | 520 | 79 | |
BT005077.1 | - | 3 - 4 | 296 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |