Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121190.3 | NP_196713.1 | 7 - 8 | 921 | 67 | |
GenBank mRNA | AY050765.1 | - | 7 - 8 | 831 | 67 | |
AY079310.1 | - | 7 - 8 | 788 | 67 | ||
U15034.1 | - | 7 - 8 | 831 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |