Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343200.1 | NP_001330242.1 | 2 - 3 | 801 | 82 | |
NM_001343201.1 | NP_001330243.1 | 2 - 3 | 523 | 82 | ||
NM_121191.4 | NP_196714.1 | 2 - 3 | 554 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |