Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KEA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121218.5 | NP_196741.2 | 10 - 11 | 1102 | 135 | |
GenBank mRNA | BT044605.1 | - | 10 - 11 | 898 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |