Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cwINV6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343222.1 | NP_001330604.1 | 4 - 5 | 1808 | 89 | |
NM_121230.3 | NP_568254.1 | 5 - 6 | 1538 | 89 | ||
GenBank mRNA | AY060553.1 | - | 5 - 6 | 1463 | 89 | |
AY141991.1 | - | 5 - 6 | 1364 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |