Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343232.1 | NP_001330601.1 | 8 - 9 | 1395 | 105 | |
NM_001343233.1 | NP_001330602.1 | 8 - 9 | 1395 | 105 | ||
NM_001343234.1 | NP_001330600.1 | 8 - 9 | 1395 | 105 | ||
NM_121267.5 | NP_568262.2 | 8 - 9 | 1395 | 105 | ||
GenBank mRNA | BT002450.1 | - | 8 - 9 | 1324 | 105 | |
BT008365.1 | - | 8 - 9 | 1240 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |