Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085107.1 | NP_001078576.1 | 8 - 9 | 1065 | 102 | |
NM_121286.4 | NP_568281.1 | 7 - 8 | 1542 | 102 | ||
GenBank mRNA | AF361634.1 | - | 7 - 8 | 1433 | 102 | |
U31836.1 | - | 7 - 8 | 1372 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |