Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335933.1 | NP_001318282.1 | 12 - 13 | 2495 | 86 | |
NM_001335934.1 | NP_001325336.1 | 13 - 14 | 2525 | 86 | ||
NM_001335935.1 | NP_001325337.1 | 9 - 10 | 2026 | 86 | ||
NM_128013.2 | NP_180028.1 | 11 - 12 | 2426 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |