Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335933.1 | NP_001318282.1 | 10 - 11 | 2161 | 81 | |
NM_001335934.1 | NP_001325336.1 | 11 - 12 | 2191 | 81 | ||
NM_001335935.1 | NP_001325337.1 | 7 - 8 | 1692 | 81 | ||
NM_128013.2 | NP_180028.1 | 9 - 10 | 2092 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |