Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AUR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124914.1 | NP_001118386.1 | 7 - 8 | 751 | 68 | |
NM_001336027.1 | NP_001325078.1 | 9 - 10 | 2042 | 68 | ||
NM_001336028.1 | NP_001325077.1 | 10 - 11 | 1898 | 68 | ||
NM_128148.4 | NP_180159.2 | 8 - 9 | 884 | 68 | ||
GenBank mRNA | AK175188.1 | - | 8 - 9 | 848 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |