Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GP ALPHA 1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336055.1 | NP_001325056.1 | 9 - 10 | 1562 | 75 | |
NM_128187.3 | NP_180198.1 | 9 - 10 | 1210 | 75 | ||
GenBank mRNA | AF385704.1 | - | 9 - 10 | 908 | 75 | |
AY093966.1 | - | 8 - 9 | 608 | 75 | ||
M32887.1 | - | 9 - 10 | 1207 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |