Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202686.2 | NP_001189615.2 | 3 - 4 | 879 | 81 | |
NM_001336110.1 | NP_001318297.1 | 4 - 5 | 944 | 81 | ||
NM_128280.3 | NP_180290.2 | 3 - 4 | 939 | 81 | ||
GenBank mRNA | AB493565.1 | - | 3 - 4 | 825 | 81 | |
DQ056543.1 | - | 3 - 4 | 825 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |