Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036353.1 | NP_001031430.1 | 4 - 5 | 882 | 71 | |
NM_128281.2 | NP_565640.1 | 4 - 5 | 845 | 71 | ||
GenBank mRNA | AB493566.1 | - | 3 - 4 | 433 | 71 | |
AJ576043.1 | - | 3 - 4 | 433 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |