Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336148.1 | NP_001318303.1 | 2 - 3 | 451 | 78 | |
NM_001336149.1 | NP_001323618.1 | 2 - 3 | 423 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |