Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UXS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084503.2 | NP_001077972.1 | 5 - 6 | 568 | 79 | |
NM_001336187.1 | NP_001325413.1 | 5 - 6 | 665 | 79 | ||
NM_128436.5 | NP_180443.1 | 5 - 6 | 695 | 79 | ||
NM_201826.3 | NP_973555.1 | 5 - 6 | 768 | 79 | ||
GenBank mRNA | AY099703.1 | - | 5 - 6 | 583 | 79 | |
AY128899.1 | - | 4 - 5 | 499 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |