Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OEP16-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128449.3 | NP_180456.1 | 2 - 3 | 369 | 89 | |
GenBank mRNA | AY045593.1 | - | 2 - 3 | 286 | 89 | |
AY093782.1 | - | 2 - 3 | 167 | 89 | ||
DQ386642.1 | - | 2 - 3 | 167 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |