Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084504.1 | NP_001077973.1 | 1 - 2 | 429 | 109 | |
NM_128462.4 | NP_180469.3 | 1 - 2 | 609 | 109 | ||
GenBank mRNA | AB195672.1 | - | 1 - 2 | 330 | 109 | |
AK228551.1 | - | 1 - 2 | 378 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |