Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTU7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128496.2 | NP_180503.1 | 1 - 2 | 353 | 76 | |
GenBank mRNA | AF288188.1 | - | 1 - 2 | 325 | 76 | |
AY045679.1 | - | 1 - 2 | 354 | 76 | ||
AY056086.1 | - | 1 - 2 | 325 | 76 | ||
AY086358.1 | - | 1 - 2 | 353 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |