Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT71C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128528.4 | NP_180535.1 | 1 | 1635 | 146 | |
GenBank mRNA | BT004155.1 | - | 1 | 1552 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |