Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113067.4 | NP_188809.2 | 3 - 4 | 1219 | 66 | |
GenBank mRNA | AK226403.1 | - | 3 - 4 | 1113 | 66 | |
AK316978.1 | - | 3 - 4 | 1111 | 66 | ||
AY140000.1 | - | 3 - 4 | 1111 | 66 | ||
BT010397.1 | - | 3 - 4 | 1039 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |