Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATXR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113077.5 | NP_188819.2 | 12 - 13 | 1374 | 79 | |
GenBank mRNA | AK221711.1 | - | 12 - 13 | 1311 | 79 | |
BT020260.1 | - | 12 - 13 | 1258 | 79 | ||
BT021112.1 | - | 12 - 13 | 1251 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |