Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATXR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113077.5 | NP_188819.2 | 9 - 10 | 1153 | 116 | |
GenBank mRNA | AK221711.1 | - | 9 - 10 | 1090 | 116 | |
BT020260.1 | - | 9 - 10 | 1037 | 116 | ||
BT021112.1 | - | 9 - 10 | 1030 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |