Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203017.2 | NP_001189946.1 | 4 - 5 | 935 | 84 | |
NM_001338571.1 | NP_001319617.1 | 4 - 5 | 997 | 84 | ||
NM_001338572.1 | NP_001325926.1 | 3 - 4 | 1163 | 84 | ||
NM_113116.4 | NP_188858.4 | 4 - 5 | 696 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |