Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338643.1 | NP_001327887.1 | 2 - 3 | 585 | 107 | |
NM_113252.4 | NP_188991.1 | 2 - 3 | 585 | 107 | ||
GenBank mRNA | AB004568.1 | - | 2 - 3 | 588 | 107 | |
AF410298.1 | - | 2 - 3 | 363 | 107 | ||
AY093708.1 | - | 2 - 3 | 348 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |