Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KINESIN-12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113271.4 | NP_189009.2 | 13 - 14 | 2837 | 76 | |
GenBank mRNA | AF540489.1 | - | 13 - 14 | 2809 | 76 | |
AY124006.1 | - | 13 - 14 | 2805 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |