Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113321.5 | NP_189058.1 | 8 - 9 | 892 | 76 | |
GenBank mRNA | AF104329.1 | - | 8 - 9 | 756 | 76 | |
AY053413.1 | - | 8 - 9 | 772 | 76 | ||
AY085022.1 | - | 8 - 9 | 792 | 76 | ||
AY133587.1 | - | 8 - 9 | 707 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |